missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-108 Monoclonal Antibody (eBio13G3-19D (13G3-19D)), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ M050T03Y03-A
This item is not returnable.
View return policy
Description
Ly108, the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6), is expressed on NK cells, B cells, and T cells. Engagement of the Ly108 receptor enhances NK cell activity and increases the expression of IFN-gamma. This receptor plays a role in the inability of NK cells to kill Epstein Barr virus-infected B cells in X-linked lymphoproliferative disease. In Ly108 knock-out mice, CD4+ T cells show a diminished ability to produce IL-4 and exhibit defective neutrophil functions, indicating the importance of Ly108 in immune regulation and cellular function. Overall, Ly108 is crucial for modulating immune responses and maintaining effective immune cell activity.
Specifications
| Ly-108 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q9ET39 | |
| SLAMF6 | |
| Affinity chromatography | |
| RUO | |
| 30925 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| IgG2a κ |
| Flow Cytometry | |
| eBio13G3-19D (13G3-19D) | |
| NovaFluor Yellow 610 | |
| SLAMF6 | |
| Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction