missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LY 2365109 hydrochloride
GREENER_CHOICE

Product Code. 18094292
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18094292 10 mg 10mg
18045772 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18094292 Supplier Tocris Bioscience™ Supplier No. 3397/10
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective GlyT1 inhibitor

Potent and selective glycine transporter 1 (GlyT1) inhibitor (IC50 values are 15.8 and > 30 000 nM at GlyT1 and GlyT2 respectively). Induces a dose-dependent elevation in CSF levels of glycine, and enhances acetylcholine and dopamine release in the striatum and prefrontal cortex respectively. Produces profound locomotor and respiratory impairments at higher doses.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868265-28-5
Molecular Formula C22H28ClNO5

Specifications

Quantity 10 mg
Formula Weight 421.91
Purity >98%
Molecular Weight (g/mol) 421.918
Chemical Name or Material LY 2365109 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.