missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-49D Rat anti-Mouse, BV750, Clone: 400000, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 747092
This item is not returnable.
View return policy
Description
Ly-49D Monoclonal antibody specifically detects Ly-49D in Mouse samples. It is validated for Flow Cytometry
Specifications
| Ly-49D | |
| Monoclonal | |
| Brilliant Violet 750 | |
| Ly49d; Ly-49d; Klra4; Chok | |
| Not Reported | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 400000 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction