missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-6A/E Recombinant Rabbit Monoclonal Antibody (24GB370)

Rabbit Recombinant Monoclonal Antibody
146.00 € - 500.00 €
Specifications
| Antigen | Ly-6A/E |
|---|---|
| Clone | 24GB370 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276247
![]() |
Invitrogen™
MA556934 |
100 μL |
500.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275751
|
Invitrogen™
MA556935 |
20 μL |
146.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ly-6A/E Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Stem Cell Antigen-1 (Sca-1) is a member of the lymphocyte antigen 6 (Ly-6) family. It is expressed on multipotent hematopoietic stem cells (HSC) and is used as a marker of HSC in mice. Sca-1 is used in combination with lineage depletion antibodies to identify murine HSC. Sca-1 can be found in fetal liver, adult bone marrow, and adult peripheral blood and spleen. Sca-1 may be involved in the regulation of B and T cell activation.Specifications
| Ly-6A/E | |
| 0.5 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05533, Q64253 | |
| 110454, 17069 | |
| A synthesized peptide derived from Mouse LY6A (aa 50-80). | |
| Antibody |
| 24GB370 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Ly6a | |
| 9804; locus A/E; Locus AE; Ly6; Ly67; Ly6a; Ly-6A.2; Ly-6A.2/Ly-6E.1; Ly6A/E; Ly-6A/E; ly6a-e; Ly6e; Ly-6E; Ly-6E.1; lymphocyte antigen 6 complex; lymphocyte antigen 6 complex, locus A; lymphocyte antigen 6 complex, locus E; lymphocyte antigen 6A-2/6E-1; Lymphocyte antigen 6E; RIG-E; Sca1; Sca-1; Sca-2; stem cell antigen 1; Stem cell antigen 2; TAP; T-cell-activating protein; thymic shared antigen 1; Tsa1; TSA-1 | |
| Ly6a, Ly6e | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title