missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6B.2 Antibody (7/4) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 13 publications
209.00 € - 499.00 €
Specifications
| Antigen | Ly-6B.2 |
|---|---|
| Clone | 44016 |
| Concentration | 1.0 mg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18071067
|
Novus Biologicals
NBP2-13077-0.025MG |
0.025 mg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18716593
|
Novus Biologicals
NBP2-13077 |
499.00 €
125µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Ly-6B.2 Monoclonal specifically detects Ly-6B.2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Ly-6B.2 | |
| 1.0 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Cultured bone marrow cells | |
| Primary | |
| Store at 4C. Do not freeze. |
| 44016 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rat | |
| Mouse | |
| IgG2a | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title