missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-6C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119794
This item is not returnable.
View return policy
Description
Positive Control: Mouse lung tissue lysate, mouse cerebellum tissue lysate, mouse lung tissue, mouse brain tissue, mouse spleen tissue, mouse heart tissue. Subcellular Location: Cell membrane.
Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
Specifications
| Ly-6C | |
| Polyclonal | |
| Unconjugated | |
| Ly6c1 | |
| AA682074; AA959465; Ly6c; Ly-6C; Ly6c1; Ly-6C1; Ly6c2; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6C1; Lymphocyte antigen Ly-6C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17067 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P0CW02 | |
| Ly6c1 | |
| Synthetic peptide within mouse Ly6c1 aa 75-100. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction