Learn More
Ly-6C Rat anti-Mouse, FITC, Clone: AL-21, BD
Description
The AL-21 antibody reacts with a non-polymorphic determinant of Ly-6C, a 14- 17-kDa GPI-linked cell-surface antigen found on some monocyte/macrophage populations, granulocytes, endothelial cells, plasma cells, and thymocyte, NK-cell, and T-subsets. Mice w
Host Species: Rat
Clone: AL-21
Isotype: IgM, κ
Species Reactivity: Mouse
Flow Cytometry
Specifications
Specifications
| Antigen | Ly-6C |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | AL-21 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Rat |
| Immunogen | Not reported |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.