missing translation for 'onlineSavingsMsg'
Learn More

LY294002, MedChemExpress

Product Code. 16433258
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433258 10 mg 10mg
16489796 50 mg 50mg
16474516 100 mg 100mg
16400926 200 mg 200mg
16461526 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16433258 Supplier Medchem Express Supplier No. HY10108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LY294002 is a broad-spectrum inhibitor of PI3K with IC50s of 0.5, 0.57, and 0.97 μM for PI3Kα, PI3Kδ and PI3Kβ, respectively. LY294002 also inhibits CK2 with an IC50 of 98 nM. LY294002 is a competitive DNA-PK inhibitor that binds reversibly to the kinase domain of DNA-PK with an IC50 of 1.4 μM. LY294002 is an apoptosis activator.

LY294002 is a broad-spectrum inhibitor of PI3K with IC50s of 0.5, 0.57, and 0.97 μM for PI3Kα, PI3Kδ and PI3Kβ, respectively. LY294002 also inhibits CK2 with an IC50 of 98 nM. LY294002 is a competitive DNA-PK inhibitor that binds reversibly to the kinase domain of DNA-PK with an IC50 of 1.4 μM. LY294002 is an apoptosis activator.

Specifications

Chemical Name or Material LY294002
CAS 154447-36-6
Color White
Purity Grade Notes Research
Molecular Formula C19H17NO3
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (162.69 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (162.69 mM; Need ultrasonic)
SMILES O=C1C=C(OC2=C1C=CC=C2C3=CC=CC=C3)N4CCOCC4
Molecular Weight (g/mol) 307.34
Formula Weight 307.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.