missing translation for 'onlineSavingsMsg'
Learn More

LY310762, MedChemExpress

Product Code. 16495728
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495728 10 mg 10mg
16411897 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495728 Supplier Medchem Express Supplier No. HY13527
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress LY310762 is a 5-HT1D receptor antagonist with Ki of 249 nM, having a weaker affinity for 5-HT1B receptor.

LY310762 is a 5-HT1D receptor antagonist with Ki of 249 nM, having a weaker affinity for 5-HT1B receptor.

Specifications

Chemical Name or Material LY310762
CAS 192927-92-7
Color White
Purity Grade Notes Research
Molecular Formula C24H28ClFN2O2
Quantity 10 mg
Solubility Information DMSO : 18.33 mg/mL (42.53 mM; Need ultrasonic)
SMILES O=C1N(CCN2CCC(C(C3=CC=C(F)C=C3)=O)CC2)C4=C(C=CC=C4)C1(C)C.[H]Cl
Molecular Weight (g/mol) 430.94
Formula Weight 430.94
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.