missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY341495, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
Description
LY341495 is a metabotropic glutamate receptor (mGluR) antagonist with IC50s of 21 nM, 14 nM, 7.8 μM, 8.2 μM, 170 nM, 990 nM, 22 μM for mGlu2, mGlu3, mGlu1a, mGlu5a, mGlu8, mGlu7, and mGlu4 receptors, respectively.
Specifications
Specifications
| Chemical Name or Material | LY341495 |
| CAS | 201943-63-7 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H19NO5 |
| Quantity | 25 mg |
| Solubility Information | DMSO : 6 mg/mL (16.98 mM; Need ultrasonic) |
| SMILES | O=C(O)[C@@H]1[C@H](C1)[C@@](N)(CC2C3=C(C=CC=C3)OC4=C2C=CC=C4)C(O)=O |
| Molecular Weight (g/mol) | 353.37 |
| Formula Weight | 353.37 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction