missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lyb-2.1 Monoclonal Antibody (CT-72.1), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517927
This item is not returnable.
View return policy
Description
Lyb-2.1 Monoclonal Antibody for Flow
CD72.1 (Lyb-2.1), a type II integral membrane glycoprotein and a member of the C-lectin family of cell surface receptors, is a differentiation antigen of B cells and is found in mouse strains expressing the Lyb-2.1 allotype. CD72.1 is the ligand of CD5 which is distributed on all T cells and a small number of B cells. The 10.1.D2 monoclonal antibody blocks binding of CD5 to CD72 on the B cell surface which leads to inhibition of the positive signal resulting from CD5/CD72 pairing. However, 10.1.D2 can itself trigger a positive signal by binding CD72.
Specifications
| Lyb-2.1 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| P21855 | |
| Cd72 | |
| Mouse CD72.1. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| CT-72.1 | |
| FITC | |
| Cd72 | |
| B-cell differentiation antigen CD72; CD72; CD72 antigen; CD72c; Ly-19; Ly32; Ly-32; Lyb-2; Ly-m19; lymphocyte antigen 32 | |
| Mouse | |
| Protein G | |
| RUO | |
| 12517 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction