missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYG2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
183.00 € - 322.00 €
Specifications
| Antigen | LYG2 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18644916
|
Novus Biologicals
NBP3-12638-50ul |
50 μg |
183.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682479
|
Novus Biologicals
NBP3-12638-100ul |
100 μg |
322.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LYG2 Polyclonal antibody specifically detects LYG2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| LYG2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| metabolism | |
| PBS, pH7.0 | |
| 254773 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.2.1.-, LYGHMGC119046, lysozyme G-like 2, lysozyme g-like protein 2, MGC119047, MGC119049 | |
| Produced in rabbits immunized with E. coli-derived Human LYG2 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title