missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lyn Antibody (LYN-01), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-519JF646
This item is not returnable.
View return policy
Description
Lyn Monoclonal specifically detects Lyn in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Lyn | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LYN | |
| Bacterially expressed recombinant fragment of human Lyn, aa 8 - 238 (NM_001111097.2). | |
| 0.1 mL | |
| Primary | |
| The LYN-01 reacts with Lyn (p56/p53), a non-receptor Src-family tyrosine kinase expressed in hematopoietic tissues. | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| LYN-01 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.10, EC 2.7.10.2, FLJ26625, JTK8, tyrosine-protein kinase Lyn, v-yes-1 Yamaguchi sarcoma viral related oncogene homolog, Yamaguchi sarcoma viral (v-yes-1) related oncogene homolog | |
| Mouse | |
| Protein A purified | |
| Protein Kinase, Signal Transduction | |
| 4067 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction