Learn More
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NBP1-97310
 View more versions of this product
Product Code. 18757513
  • 493.00 € / 0.10mL
Description
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Polyclonal specifically detects Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B
Polyclonal
Unconjugated
PBS (pH 7.4) with 0.02% Sodium Azide
CT31FLJ10538, EC 1.14.11, EC 1.14.11.-, homolog 1A, JARID1BFLJ16281, jumonji, AT rich interactive domain 1B, Jumonji, AT rich interactive domain 1B (RBP2-like), lysine (K)-specific demethylase 5B, PLU-1FLJ12459, PLU1FLJ23670, PUT1, RBBP2H1, RBBP2H1AFLJ12491, RBP2-H1, Retinoblastoma-binding protein 2 homolog 1
Rabbit
Affinity Purified
RUO
Primary
Human
IgG
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.828 mg/mL
Western Blot reported in scientific literature (PMID 23354547), Flow Cytometry, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200
Q9UGL1
KDM5B
A synthetic peptide made to an internal portion of the human JARID1B protein (between residues 800-850) [UniProt Q9UGL1]
0.1 mL
Cancer, Epigenetics
10765
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.