Learn More
Invitrogen™ LysoLight™ Antibody Labeling Kits

Description
LysoLight Antibody Labeling Kits provide a powerful tool for visualizing the lysosomal localization and catabolic degradation of antibodies and proteins. These kits utilize a unique fluorophore that is only active upon enzymatic cleavage by the lysosomal specific enzyme, Cathepsin B. Outside of the lysosome, LysoLight dyes have little to no observable background fluorescence, and only upon localization to the protease rich environment of the lysosome, does LysoLight become activated and brightly fluoresce bright green or deep red in color.
Spécification
Spécification
| Color | Deep Red |
| Description | LysoLight™ Deep Red Antibody Labeling Kit |
| Detection Method | Fluorescence |
| For Use With (Application) | Antibody and Protein Labeling |
| For Use With (Equipment) | Flow Cytometer, Fluorescence Microscope, High Content Analysis Instrument |
| Product Type | Antibody Labeling Kit |
| Product Line | LysoLight |
| Quantity | 3 x 100 μg |
| Solubility | DMSO (Dimethylsulfoxide) |
| Excitation/Emission | 650/668 nm |
For Research Use Only.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.