missing translation for 'onlineSavingsMsg'
Learn More

m-Anisidine, 99%

Product Code. 15710789 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
Unit Size:
1 L
100mL
250mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15710789 250 mL 250mL
10335880 5 mL 5mL
10606482 100 mL 100mL
10637354 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15710789 Supplier Thermo Scientific Acros Supplier No. 104822500
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 536-90-3 | C7H9NO | 123.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-90-3
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007783
InChI Key NCBZRJODKRCREW-UHFFFAOYSA-N
Synonym m-anisidine, 3-aminoanisole, m-methoxyaniline, 3-methoxybenzenamine, 3-anisidine, m-aminoanisole, benzenamine, 3-methoxy, m-anisylamine, 1-amino-3-methoxybenzene, unii-jxa144kx2i
PubChem CID 10824
IUPAC Name 3-methoxyaniline
SMILES COC1=CC=CC(=C1)N

Specifications

Color Yellow to Brown
Density 1.1020g/mL
Boiling Point 250°C to 251°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4NH2
Refractive Index 1.5800 to 1.582
Quantity 250 mL
Beilstein 13,404
Merck Index 14,667
Specific Gravity 1.102
Solubility Information Solubility in water: 18g/L (20°C). Other solubilities: soluble in acids,soluble in alcohol,diethyl ether,acetone and,benzene
Formula Weight 123.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Anisidine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.