missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol, 0.99%, Honeywell™

Product Code. 15677780 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
2 kg
Packaging:
Glass bottle
Unit Size:
100g
250g
2kg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15677780 5 g Glass bottle 5g
15647780 100 g Glass bottle 100g
15657780 250 g Glass bottle 250g
15667780 2 kg Glass bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15677780 Supplier Honeywell Research Chemicals Supplier No. C857275G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

0.99

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002302
InChI Key RLSSMJSEOOYNOY-UHFFFAOYSA-N
Synonym m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol
PubChem CID 342
ChEBI CHEBI:17231
IUPAC Name 3-methylphenol
SMILES CC1=CC(=CC=C1)O

Specifications

Melting Point 11.8°C (1013hPa)
Color Colorless to Yellow
Density 1.034g/mL at 25°C
Boiling Point 202°C (1013 hPa)
Flash Point 86°C (186.8°F)
Ignition Point 1036°F
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3C6H4OH
Quantity 5 g
UN Number UN2076
Beilstein 506719
Solubility Information 22.7g/L at 25°C at ca. 7 pH Solubility in water
Vapor Density 3.72 (vs air)
Formula Weight 108.14g/mol
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material m-Cresol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.