missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M-CSFR/CD115 Antibody (CL11653) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44137
This item is not returnable.
View return policy
Description
M-CSFR/CD115 Monoclonal antibody specifically detects M-CSFR/CD115 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| M-CSFR/CD115 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry/ Immunofluorescence 2-10 μg/mL | |
| CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms | |
| This antibody was generated using a recombinant protein sequence of P07333, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase | |
| 1436 | |
| Human | |
| Purified |
| Immunocytochemistry/Immunofluorescence | |
| CL11653 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction