missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M-Ras/R-Ras3 Antibody (S07-2A5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19680-100ul
This item is not returnable.
View return policy
Description
M-Ras/R-Ras3 Monoclonal antibody specifically detects M-Ras/R-Ras3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| M-Ras/R-Ras3 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| S07-2A5 | |
| Unconjugated | |
| FLJ42964, muscle and microspikes RAS, muscle RAS oncogene homolog, ras-related protein M-Ras, Ras-related protein R-Ras3, R-RAS3, RRAS3M-RAs | |
| A synthetic peptide of human M-Ras/R-Ras3 (Uniprot # O14807) | |
| 100 μg | |
| Signal Transduction | |
| 22808 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction