missing translation for 'onlineSavingsMsg'
Learn More

m-Toluidine, 99%

Product Code. 11437976
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437976 1000 g 1000g
11447976 250 g 250g
11457976 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11437976 Supplier Thermo Scientific Alfa Aesar Supplier No. A14058.0B
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 108-44-1 | C7H9N | 107.16 g/mol

m-Toluidine is used in the manufacture of organic chemicals. It is also used in manufacture of various dyes and pigments, photo chemicals, pharmaceutical intermediates.antioxidants, curing agent, accelerator for polymer and rubber industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Toluidine is used in the manufacture of organic chemicals. It is also used in manufacture of various dyes and pigments, photo chemicals, pharmaceutical intermediates.antioxidants, curing agent, accelerator for polymer and rubber industry.

Solubility
Soluble in water (2g/L), (10 g/L at 25°C ).

Notes
Keep container tightly sealed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-44-1
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00007808
InChI Key JJYPMNFTHPTTDI-UHFFFAOYSA-N
Synonym m-toluidine, 3-toluidine, benzenamine, 3-methyl, 3-aminotoluene, m-methylaniline, m-tolylamine, m-toluidin, m-aminotoluene, m-methylbenzenamine, 3-methylbenzenamine
PubChem CID 7934
IUPAC Name 3-methylaniline
SMILES CC1=CC=CC(N)=C1

Specifications

Melting Point -30°C
Density 0.992
Boiling Point 203°C to 204°C
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.567
Quantity 1000 g
UN Number UN1708
Beilstein 635944
Merck Index 14,9536
Solubility Information Soluble in water (2g/L),(10g/L at 25°C ).
Formula Weight 107.16
Percent Purity 99%
Chemical Name or Material m-Toluidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.