missing translation for 'onlineSavingsMsg'
Learn More

m-Toluidine, 99%

Product Code. 10307670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
1 L
5 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
5 L
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10307670 5 mL Glass Bottle 5mL
10626172 1 L Glass Bottle 1 L
15109203 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10307670 Supplier Thermo Scientific Acros Supplier No. 157870050
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-44-1 | C7H9N | 107.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-44-1
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00007808
InChI Key JJYPMNFTHPTTDI-UHFFFAOYSA-N
Synonym m-toluidine, 3-toluidine, benzenamine, 3-methyl, 3-aminotoluene, m-methylaniline, m-tolylamine, m-toluidin, m-aminotoluene, m-methylbenzenamine, 3-methylbenzenamine
PubChem CID 7934
IUPAC Name 3-methylaniline
SMILES CC1=CC=CC(N)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -30.0°C
Color White
Density 0.9800g/mL
Boiling Point 203.0°C to 204.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3C6H4NH2
Refractive Index 1.5660 to 1.5680
Quantity 5 mL
Beilstein 12,853
Merck Index 15,9696
Specific Gravity 0.98
Solubility Information Solubility in water: 0.2 g/100mL (20°C). Other solubilities: >50 g/100mL methanol, >50 g/100mL ethanol, soluble in ether, dmf and diluted acids
Formula Weight 107.15
Percent Purity 99%
Physical Form Crystalline Solid or Liquid
Chemical Name or Material m-Toluidine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.