missing translation for 'onlineSavingsMsg'
Learn More

M-Xylene, Pure, Fisher Chemical

Product Code. 10182350 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10182350 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10182350 Supplier Fisher Chemical Supplier No. X/0350/08
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Specifications

Packaging Amber glass bottle
CAS 108-38-3
Vapor Pressure 8mbar at 20°C
Viscosity 0.62 mPaS at 20°C
Vapor Density &3.66
Physical Form Liquid
Molecular Formula C8H10
MDL Number MFCD00008536
Color Colorless
Melting Point -48°C
Boiling Point 139°C
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILES CC1=CC(C)=CC=C1
IUPAC Name 1,3-xylene
Molecular Weight (g/mol) 106.17
PubChem CID 7929
Quantity 500 mL
ChEBI CHEBI:28488
Formula Weight 106.17g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • m-Xylene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.