missing translation for 'onlineSavingsMsg'
Learn More

m-Xylylene dibromide, 97%

Product Code. 11471497
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471497 10 g 10g
11481497 50 g 50g
11491497 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471497 Supplier Thermo Scientific Alfa Aesar Supplier No. B24959.09
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 626-15-3 | C8H8Br2 | 263.96 g/mol

It is used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macro cyclic salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macro cyclic salt.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, active metals, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-15-3
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000178
InChI Key OXHOPZLBSSTTBU-UHFFFAOYSA-N
Synonym 1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene
PubChem CID 69373
IUPAC Name 1,3-bis(bromomethyl)benzene
SMILES C1=CC(=CC(=C1)CBr)CBr

Specifications

Melting Point 74°C to 76°C
Quantity 10 g
UN Number UN3261
Beilstein 971085
Solubility Information Insoluble in water.
Formula Weight 263.96
Percent Purity 97%
Chemical Name or Material m-Xylylene dibromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.