missing translation for 'onlineSavingsMsg'
Learn More

m-Xylylenediamine, 99%

Product Code. 10741925 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10741925 500 mL Glass bottle 500mL
10285140 5 mL Glass bottle 5mL
10627112 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10741925 Supplier Thermo Scientific Acros Supplier No. 141025000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1477-55-0 | C8H12N2 | 136.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1477-55-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.2
MDL Number MFCD00008119
InChI Key FDLQZKYLHJJBHD-UHFFFAOYSA-N
Synonym m-xylylenediamine, 1,3-benzenedimethanamine, 1,3-bis aminomethyl benzene, 1,3-xylylenediamine, 1,3-phenylenedimethanamine, m-xylylendiamin, 1,3-xylenediamine, mxda, m-phenylenebis methylamine, methylamine, m-phenylenebis
PubChem CID 15133
IUPAC Name [3-(aminomethyl)phenyl]methanamine
SMILES C1=CC(=CC(=C1)CN)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.0300g/mL
Boiling Point 265°C (745.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H4(CH2NH2)2
Refractive Index 1.5700 to 1.572
Quantity 500 mL
Beilstein 13, IV, 293
Specific Gravity 1.03
Formula Weight 136.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Xylylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.