missing translation for 'onlineSavingsMsg'
Learn More

M344, MedChemExpress

Product Code. 16471877
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471877 50 mg 50mg
16468407 5 mg 5mg
16445718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16471877 Supplier Medchem Express Supplier No. HY13506
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress M344 (D 237) is an inhibitor of histone deacetylase (IC50=100 nM) and an inducer of terminal cell fifferentiation.

M344 (D 237) is an inhibitor of histone deacetylase (IC50=100 nM) and an inducer of terminal cell fifferentiation.

Specifications

Chemical Name or Material M344
Physical Form Solid
CAS 251456-60-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H25N3O3
For Use With (Application) Cancer-programmed cell death
Synonym D 237 MS 344
Solubility Information DMSO : ≥ 100 mg/mL (325.32 mM)
SMILES O=C(NCCCCCCC(NO)=O)C1=CC=C(N(C)C)C=C1
Molecular Weight (g/mol) 307.39
Formula Weight 307.39
Percent Purity 99.28%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.