missing translation for 'onlineSavingsMsg'
Learn More

MA5+Mo in 75 cSt Hydrocarbon Oil

Product Code. 30316796
Change view
Click to view available options
Quantity:
400 g
Unit Size:
400g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316796 400 g 400g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316796 Supplier VHG Labs Supplier No. VHGMA5+MO1500400G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium∣Calcium∣Magnesium∣Molybdenum∣Phosphorus∣Zinc
CAS 7440-39-3, 7440-70-2, 7439-95-4, 7439-98-7, 7723-14-0, 7440-66-6
Analyte or Component Names Barium∣Calcium∣Magnesium∣Molybdenum∣Phosphorus∣Zinc
Shipping Condition Room Temperature
Solution Type Mixture
Element Multi-element
Solvent or Matrix 75 cSt Hydrocarbon Oil
Industry Type Industrial QC
Quantity 400 g
Concentration or Composition (by Analyte or Components) Barium 1500 μg/g∣Calcium 1500 μg/g∣Magnesium 1500 μg/g∣Molybdenum 1500 μg/g∣Phosphorus 1500 μg/g∣Zinc 1500 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.