missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ Reversed Phase HPLC Columns

Product Code. 15407162 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
Diameter (Metric):
1 mm
2.1 mm
3 mm
Stationary Phase:
PSDVB (Polystyrene Divinylbenzene)
Phenyl (Ph)
Unit Size:
Each
Pack of 2
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Stationary Phase unitSize
15407162 100 mm 3 mm Phenyl (Ph) Each
15417162 50 mm 3 mm Phenyl (Ph) Each
15427162 10 mm 3 mm Phenyl (Ph) Pack of 2
15308539 100 mm 2.1 mm Phenyl (Ph) Each
15437162 50 mm 2.1 mm Phenyl (Ph) Each
15447162 10 mm 2.1 mm Phenyl (Ph) Pack of 2
15776222 50 mm 1 mm Phenyl (Ph) Each
16853740 50 mm 1 mm PSDVB (Polystyrene Divinylbenzene) Each
16864106 100 mm 1 mm PSDVB (Polystyrene Divinylbenzene) Each
16874106 150 mm 1 mm PSDVB (Polystyrene Divinylbenzene) Each
16307045 150 mm 2.1 mm - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 15407162 Supplier Thermo Scientific™ Supplier No. 088644
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.

Use the new Thermo Scientific™ MAbPac™ Reversed Phase columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.

Compatible with organic solvent used in HPLC and MS under RP conditions
  • Resin is based on a hydrophobic supermacroporous 4μm polymer particles
  • Stable at extreme pH (0 to 14) and high temperature (up to 110 °C)
Efficient with low carry over
  • 1,500Å large pore size polymeric resin
  • Efficient separation of protein molecules with very low carry over
Designed for reversed-phase biomolecule LC and LC/MS separations
  • Superior resolution power for monoclonal antibodies and related substances
  • High efficiency with low carry-over
  • Excellent MS compatibility
  • Wide operating pH range: 0 to 14
  • High temperature stability: up to 110°C
  • High throughput
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 3 mm
Length (Metric) 100 mm
Max. Pressure 4000 psi (275 bar)
Particle Size 4 μm
pH 0 to 14
Pore Size 1500 Å
Quantity Each
Temperature <110°C
Stationary Phase Phenyl (Ph)
Product Line MAbPac
Packing Material DVB (Divinylbenzene) Polymer
Column Type Reversed Phase
Phase Reversed Phase
Technique Bio-LC
Green Features FSC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.