Learn More
MACC1 Antibody (CL0856) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52955AF350
Product Code. 30290728
  • 737.00 € / 0.10mL
Description
MACC1 Monoclonal antibody specifically detects MACC1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MACC1
Monoclonal
Alexa Fluor 350
metastasis associated in colon cancer 1, SH3 domain-containing protein 7a5,7A5
This antibody was developed using a recombinant protein derived from Q6ZN28, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
346389
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL0856
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified