missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macro H2A.2 Antibody (CL5263) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61156H
This item is not returnable.
View return policy
Description
Macro H2A.2 Monoclonal antibody specifically detects Macro H2A.2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Macro H2A.2 | |
| Monoclonal | |
| HRP | |
| core histone macro-H2A.2, core histone macroH2A2.2, H2A histone family, member Y2, Histone macroH2A2, macroH2A2, mH2A2 | |
| This antibody was developed using a synthetic peptide derived from Q9P0M6, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL5263 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 55506 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction