missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macrophage Rat anti-Mouse, Clone: MOMA-2, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-64946-0.05MG
This item is not returnable.
View return policy
Description
Macrophage Monoclonal specifically detects Macrophage in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Knockout Validated.
Specifications
| Macrophage | |
| Monoclonal | |
| 0.5 mg/ml | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), KnockDown | |
| MOMA-2 | |
| Unconjugated | |
| EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
| Mouse lymph node stroma | |
| 0.05 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction