missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Macrophage-SFM (1X)

Product Code. 11500426 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11500426 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11500426 Supplier Gibco™ Supplier No. 12065074
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Macrophage-SFM (1X)

Macrophage-SFM is a serum-free medium designed for the culture of human peripheral blood monocytes and macrophages (1).

Contains L-glutamine, but no antibiotics.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Macrophages, Monocytes
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Sterile-filtered
With Additives Glutamine
Quantity 500 mL
Shipping Condition Room Temperature
Endotoxin Level ≤1 EU/mL, ≤ 1 EU/mL
Included Antibiotics No antibiotics
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.