missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAD1 Recombinant Rabbit Monoclonal Antibody (24GB3620)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557079
This item is not returnable.
View return policy
Description
MAD1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
Specifications
| MAD1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MAD1L1 | |
| AI173502; AW550425; coiled-coil domain-containing protein MAD1; hMAD1; HsMAD1; Mad1; MAD1 mitotic arrest deficient 1-like 1; MAD1 mitotic arrest deficient like 1; MAD1 mitotic arrest deficient-like 1; MAD1 mitotic arrest deficient-like 1 (yeast); MAD1L1; MAD1-like protein 1; mitotic arrest deficient 1-like 1; mitotic arrest deficient 1-like protein 1; Mitotic arrest deficient protein 1; Mitotic checkpoint MAD1 protein homolog; mitotic spindle assembly checkpoint protein MAD1; mitotic-arrest deficient 1, yeast, homolog-like 1; MXD1; PIG9; Spindle assembly checkpoint component MAD1; Tax-binding protein 181; TP53I9; tumor protein p53 inducible protein 9; TXBP181; YGL086W | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17120, 680006, 8379 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3620 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9WTX8, Q9Y6D9 | |
| MAD1L1 | |
| A synthesized peptide derived from human MAD1 (aa 50-170). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction