missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MADD Antibody (SR1358), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22550-100ul
This item is not returnable.
View return policy
Description
MADD Monoclonal antibody specifically detects MADD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MADD | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1358 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| DENNFLJ36300, Differentially expressed in normal and neoplastic cells, FLJ35600, IG20, Insulinoma glucagonoma clone 20, insulinoma-glucagonoma protein 20, KIAA0358RAB3GEP, MAP kinase-activating death domain protein, MAP-kinase activating death domain, Rab3 GDP/GTP exchange factor | |
| A synthesized peptide derived from human MADD (Uniprot #: Q8WXG6) | |
| 100 μg | |
| Signal Transduction | |
| 8567 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction