missing translation for 'onlineSavingsMsg'
Learn More

Mafenide Acetate, MedChemExpress

Product Code. 16494417
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16494417

Brand: Medchem Express HYB0614A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mafenide Acetate is an effective sulfonamide-type antimicrobial agent used for burn wounds. Mafenide Acetate shows activity against both Gram-positive and Gram-negative organisms, including Pseudomonas aeruginosa, via inhibition of nucleotide synthesis.

Mafenide Acetate is an effective sulfonamide-type antimicrobial agent used for burn wounds. Mafenide Acetate shows activity against both Gram-positive and Gram-negative organisms, including Pseudomonas aeruginosa, via inhibition of nucleotide synthesis.

Specifications

Chemical Name or Material Mafenide Acetate
Physical Form Solid
CAS 13009-99-9
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C9H14N2O4S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (406.04 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (406.04 mM)
SMILES O=S(C1=CC=C(CN)C=C1)(N)=O.CC(O)=O
Molecular Weight (g/mol) 246.28
Formula Weight 246.28
Percent Purity 99.43%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.