missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00 € - 526.00 €
Specifications
| Antigen | MAGEA4 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694818
|
Novus Biologicals
NBP2-41318-0.025mg |
0.025 mg |
168.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18059153
|
Novus Biologicals
NBP2-41318 |
0.1 mg |
526.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAGEA4 Polyclonal specifically detects MAGEA4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence, Immunocytochemistry.Specifications
| MAGEA4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| MAGE-X2 antigen, melanoma antigen family A, 4, melanoma-associated antigen 4, member 4 | |
| MAGEA4 | |
| IgG | |
| Affinity Purified | |
| 45 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| PBS with 0.02% Sodium Azide | |
| 4103 | |
| MAGEA4 antibody was raised against a 16 amino acid peptide near the amino terminus of human MAGEA4. The immunogen is located within the first 50 amino acids of MAGEA4. Amino Acid Squence: VGAQAPTTEEQEAAVS | |
| Primary | |
| MAGEA4 antibody is human specific. At least four isoforms of MAGEA4 are known to exist. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title