missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB4 Antibody (OTI3H2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00 € - 550.00 €
Specifications
| Antigen | MAGEB4 |
|---|---|
| Clone | OTI3H2 |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:500 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306220
|
Novus Biologicals
NBP3-44915-100ul |
100 μL |
550.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306367
|
Novus Biologicals
NBP3-44915-30ul |
30 μL |
256.00 €
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAGEB4 Monoclonal antibody specifically detects MAGEB4 in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| MAGEB4 | |
| Western Blot 1:2000, Immunohistochemistry 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 4115 | |
| IgG2b | |
| >95% | |
| Antibody |
| OTI3H2 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| cancer/testis antigen family 3, member 6, CT3.6, MAGE-B4 antigen, melanoma antigen family B, 4, melanoma-associated antigen B4, MGC33144 | |
| Human recombinant protein fragment corresponding to amino acids 1-193 of human MAGEB4(NP_002358) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title