missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAGEC2 Recombinant Rabbit Monoclonal Antibody (K01_1L17)

Product Code. 30824625
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30824625 100 μL
30830407 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30824625 Supplier Invitrogen™ Supplier No. MA563206

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MAGEC2 Recombinant Monoclonal Antibody for IHC (P), Western Blot

MAGE-C2/C3 (Melanoma-associated antigen C2, Hepatocellular carcinoma-associated antigen 587) is a 373 amino acid protein encoded by the human gene MAGEC2. Genes of the MAGE family direct the expression of tumor antigens that are recognized on human melanomas by autologous cytolytic T lymphocytes. Among tumor tissues, MAGE-C2/C3 is frequently expressed in seminomas, melanomas, and bladder transitional cell carcinomas. It is also expressed in a significant fraction of head and neck carcinomas, breast carcinomas, nonsmall-cell lung carcinomas, and sarcomas. In hepatocellular carcinoma, there is a inverse correlation between tumor differentiation and protein expression, i. e. the lower the differentiation, the higher percentage of expression. MAGE-C2/C3 is not expressed in normal tissues, except in germ cells in the seminiferous tubules and in Purkinje cells of the cerebellum. It is strongly expressed in spermatogonia and primary spermatocytes, however, at later stages of maturation, expression gradually decreases to become undetectable in mature spermatids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1L17
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene MAGEC2
Gene Accession No. Q9UBF1
Gene Alias Cancer/testis antigen 10; CT10; HCA587; hepatocellular cancer antigen 587; hepatocellular carcinoma-associated antigen 587; MAGE family member C2; MAGEC2; MAGE-C2; MAGE-C2 antigen; MAGEE1; MAGE-E1 antigen; melanoma antigen family C, 2; melanoma antigen family C2; melanoma antigen, family E, 1, cancer/testis specific; melanoma-associated antigen C2; uncharacterized protein LOC317590
Gene Symbols MAGEC2
Host Species Rabbit
Immunogen Recombinant protein of human MAGEE1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51438
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.