missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MAGI3 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30984-50ug
This item is not returnable.
View return policy
Description
MAGI3 Polyclonal antibody specifically detects MAGI3 in Human samples. It is validated for Western Blot
Specifications
| MAGI3 | |
| Polyclonal | |
| Western Blot : 1-10 μg/mL | |
| dJ730K3.2, MAGI-3, membrane associated guanylate kinase, WW and PDZ domain containing 3, membrane-associated guanylate kinase inverted 3, membrane-associated guanylate kinase-related 3, WW and PDZ domain-containing protein 3 | |
| Recombinant protein corresponding to PDZ domains 4-5 of MAGI3 | |
| 50 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 260425 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur