Learn More
Fisherbrand™ MAGic Clamp™ Universal Platform
Product Code. 16363226 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Mini Incubator 16313226
Shaking Incubator 16333226
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16363226 Mini Incubator 16313226 Each
16393246 Shaking Incubator 16333226 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16363226
  • ONLINE EXCLUSIVE PRICE
    345.00 €
    256.13 € / Pack of 1
    Save 88.87 €
    25% Off
Estimated Shipment: 05-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fisherbrand™ MAGic Clamp™ Universal Platform features a unique magnetic attachment designed to make changing flask clamps instant and easy.
  • For use with shaking incubator
  • Magnetic
  • Can hold a wide variety of clamps for tubes and flasks
Specifications
Product Type Universal Platform
Material Steel
Dimensions (L x W) 9.5 x 11.5 cm (3.74 x 4.53 in.)
Description Magnetic Attachment
For Use With (Application) Holding racks and clamps
For Use With (Equipment) Mini Incubator 16313226