missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hexafluorosilicate hexahydrate, 98%

Product Code. 11444594
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
This item is not returnable. View return policy

Product Code. 11444594

Brand: Thermo Scientific Alfa Aesar A13189.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18972-56-0 | MgSiF6·6H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18972-56-0
Molecular Formula MgSiF6·6H2O
MDL Number MFCD00150250

Specifications

Quantity 500 g
UN Number UN2853
Formula Weight 274.47 (166.39 Anhydrous)
Percent Purity 98%
Odor Odorless
Density 1.79
Chemical Name or Material Magnesium hexafluorosilicate hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.