missing translation for 'onlineSavingsMsg'
Learn More

Magnesium solution for ICP 10g/L HNO3 matrix, Fisher chemical™
BIOPHARMA

Product Code. 12986334 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12986334 500 mL Bottle 500mL
12966334 100 mL Bottle 100mL
12976334 250 mL Bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12986334 Supplier Fisher Chemical Supplier No. S 232 500
  • ONLINE EXCLUSIVE PRICE
    251.00 €
    248.48 € / 500mL
    Save 2.52 €
    1% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
Analyte or Component Names Mg
Concentration 10±0.02 g/L
Type Mass Spectrometry Standard
Element Mg
Solvent or Matrix Nitric Acid
Quantity 500 mL
compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. Magnesium (Mg) 10000mg/l in HNO3 2%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.