missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Pack of 16 stainless steel bag mounting strips
- Use to attach vessels to the magnetic platform of the Roto-Shake Genie, Rotator Genie, Enviro-Genie, or Incubator-Genie
Specifications
Specifications
| Product Type | Magnetic Mounting Strips |
| Material | Stainless Steel |
| Color | Silver |
| For Use With (Equipment) | Roto-Shake Genie, Rotator Genie, Enviro-Genie or Incubator-Genie |
| Packaging Quantity | 16 Strips |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?