missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGP-1/MFAP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 529.00 €
Specifications
| Antigen | MAGP-1/MFAP2 |
|---|---|
| Concentration | 0.1mg/mL |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18412682
|
Novus Biologicals
NBP1-87735-25ul |
25 μL |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18213666
|
Novus Biologicals
NBP1-87735 |
0.1 mL |
529.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAGP-1/MFAP2 Polyclonal specifically detects MAGP-1/MFAP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MAGP-1/MFAP2 | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P55001 | |
| 4237 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QYTHYSDQIDNPDYYDYQEVTPRPSEEQFQFQSQQQVQQEVIPAPTPEPGNAELEPTEPGPLDCREEQYPCTRLYSIHRPCKQCLNEVCFYSLRRVYVINKEICVRTVCAHEELLRADLCRDKFSKCG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.1mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MAGP1, MAGP-1FLJ50901, MAGPMFAP-2, Microfibril-associated glycoprotein 1, microfibrillar-associated protein 2 | |
| MFAP2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title