Learn More
Major Basic Protein (PRG2) Monoclonal Antibody (OTI1C12), OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA813063S
Encompass_Preferred
Product Code. 17507734
  • 242.00 € / 30µL
Description
Major Basic Protein (PRG2) Monoclonal antibody specifically detects Major Basic Protein (PRG2) in Human samples. It is validated for Western Blot
Specifications
Specifications
Major Basic Protein (PRG2)
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P13727
Mouse
Affinity chromatography
RUO
5553
-20°C
IgG2a
Western Blot
OTI1C12
Unconjugated
PRG2
BMPG; bone marrow proteoglycan; bone-marrow proteoglycan; EMBP; Eosinophil granule major basic protein; eosinophil major basic protein; LPR3; major basic protein 1; MBP; MBP1; Mbp-1; mMBP; mMBP-1; natural killer cell activator; Pregnancy-associated major basic protein; PRG2; PRG-2; proMBP; Proteoglycan 2; proteoglycan 2 preproprotein; proteoglycan 2, bone marrow; proteoglycan 2, bone marrow (natural killer cell activator, eosinophil granule major basic protein); proteoglycan 2, pro eosinophil major basic protein
Human recombinant protein fragment corresponding to amino acids 17-222 of human PRG2 (NP_002719) produced in E.coli.
30 μL