missing translation for 'onlineSavingsMsg'
Learn More

Major Environmental Elements: Na, Ca, K, Mg @ 1000 μg/mL in 2% HNO3, VHG™

Product Code. 17960866
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17960866 100 mL 100mL
17952856 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17960866 Supplier VHG Labs Supplier No. VHGSM41000100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Major Environmental Elements: Na, Ca, K, Mg @ 1000 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium/Magnesium/Potassium/Sodium
CAS 7440-70-2, 7439-95-4, 9-7-7440, 7440-23-5
Methods ICP/ICPMS
Analyte or Component Names Calcium/Magnesium/Potassium/Sodium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 2% HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Calcium 1000 μg/mL/Magnesium 1000 μg/mL/Potassium 1000 μg/mL/Sodium 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.