missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MALT1 Antibody (SR2292), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22210-50ul
This item is not returnable.
View return policy
Description
MALT1 Monoclonal antibody specifically detects MALT1 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| MALT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR2292 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| caspase-like protein, DKFZp434L132, EC 3.4.22.-, MALT associated translocation, MALT lymphoma-associated translocation, MLT1, MLTMALT-lymphoma associated translocation, mucosa associated lymphoid tissue lymphoma translocation gene 1, mucosa-associated lymphoid tissue lymphoma translocation protein 1, paracaspase | |
| A synthesized peptide derived from human MALT1 (Uniprot #: Q9UDY8) | |
| 50 μg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 10892 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction