missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Maltodextrin 12

Product Code. 16661824 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16661824 50 g 50g
16671824 250 g 250g
16651824 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16661824 Supplier Thermo Scientific Chemicals Supplier No. 466680500
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
This item is not returnable. View return policy

CAS: 9050-36-6 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9050-36-6
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O

Specifications

Melting Point 240.0°C
Quantity 50 g
Chemical Name or Material Maltodextrin 12

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.