missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAMLD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
183.00 € - 322.00 €
Specifications
| Antigen | MAMLD1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18657527
|
Novus Biologicals
NBP3-12581-100ul |
100 μg |
322.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603497
|
Novus Biologicals
NBP3-12581-50ul |
50 μg |
183.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAMLD1 Polyclonal antibody specifically detects MAMLD1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| MAMLD1 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| chromosome X open reading frame 6, CXorf6Protein CG1, F18CG1mastermind-like domain-containing protein 1, HYSP2, mastermind-like domain containing 1 | |
| Produced in rabbits immunized with E. coli-derived Human MAMLD1 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 10046 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title