missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences pCSG-IBA Mammalian Expression Vector with Secretion
Designed for high-level episomal expression in mammalian hosts under G418 selection. Mammalian Expression Vector with Secretion is a large expression vector with universal features for transient expression as well as for generation of stable cell lines (NeoR).
266.00 €
Specifications
Constitutive or Inducible System | Constitutive |
---|---|
Marker Gene | Ampicillin |
Promoter | CMV |
Product Type | Vector |
Selection Agent (Eukaryotic) | Geneticin™ (G-418) |
Product Code | Brand | Protein Tag | Vector | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Protein Tag | Vector | Price | Quantity & Availability | ||||
15603858
|
IBA Lifesciences
5-5001-001 |
Untagged | pCSG-IBAwt2 |
266.00 €
5µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15603878
|
IBA Lifesciences
5-5144-001 |
Twin-Strep-tag, His Tag (6x) | pCSG-IBA144 |
266.00 €
5µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15633868
|
IBA Lifesciences
5-5102-001 |
Twin-Strep-tag | pCSG-IBA102 |
266.00 €
5µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15653868
|
IBA Lifesciences
5-5104-001 |
Twin-Strep-tag | pCSG-IBA104 |
266.00 €
5µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15683868
|
IBA Lifesciences
5-5142-001 |
His Tag (6x), Twin-Strep-tag | pCSG-IBA142 |
266.00 €
5µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Designed for high-level episomal expression in mammalian hosts under G418 selection. IBA Lifesciences GmbH Mammalian Expression Vector with Secretion is a large expression vector with universal features for transient expression as well as for generation of stable cell lines (NeoR).Specifications
Constitutive | |
CMV | |
Geneticin™ (G-418) | |
5 μg |
Ampicillin | |
Vector | |
Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage | |
Episomal expression (oriP, EBNA-1); BM40 secretion signal |