missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAN1A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97194-50ul
This item is not returnable.
View return policy
Description
MAN1A2 Polyclonal antibody specifically detects MAN1A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MAN1A2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Alpha-1,2-mannosidase IB, EC 3.2.1.113, MAN1Balpha1,2-mannosidase, Mannosidase alpha class 1A member 2, mannosidase, alpha, class 1A, member 2, mannosyl-oligosaccharide 1,2-alpha-mannosidase IB, Processing alpha-1,2-mannosidase IB | |
| Produced in rabbits immunized with E. coli-derived Human MAN1A2 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10905 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction